Assay Details
Gene Symbol: cnih2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013524.1 | NP_001013542.1 | 5 - 6 | 756 | 91 | |
XM_021479171.1 | XP_021334846.1 | 7 - 8 | 1313 | 91 | ||
XM_021479172.1 | XP_021334847.1 | 7 - 8 | 1310 | 91 | ||
GenBank mRNA | BC090830.1 | - | 5 - 6 | 756 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
