Assay Details
Gene Symbol: henmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017842.2 | NP_001017842.1 | 3 - 4 | 440 | 95 | |
XM_009294718.3 | XP_009292993.1 | 3 - 4 | 1295 | 95 | ||
GenBank mRNA | BC092772.1 | - | 3 - 4 | 445 | 95 | |
BC165189.1 | - | 3 - 4 | 376 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
