Assay Details
Gene Symbol: mtmr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020721.1 | NP_001018557.1 | 12 - 13 | 1612 | 64 | |
XM_005172285.4 | XP_005172342.1 | 13 - 14 | 1751 | 64 | ||
XM_005172286.4 | XP_005172343.1 | 12 - 13 | 1685 | 64 | ||
XM_009303059.3 | XP_009301334.1 | 14 - 15 | 1775 | 64 | ||
XM_009303060.3 | XP_009301335.1 | 13 - 14 | 1709 | 64 | ||
GenBank mRNA | BC095884.1 | - | 12 - 13 | 1612 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
