Assay Details
Gene Symbol: mtmr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020721.1 | NP_001018557.1 | 2 - 3 | 344 | 63 | |
GenBank mRNA | BC095884.1 | - | 2 - 3 | 344 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
