Assay Details
Gene Symbol: mtmr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020721.1 | NP_001018557.1 | 4 - 5 | 488 | 67 | |
XM_005172285.4 | XP_005172342.1 | 5 - 6 | 627 | 67 | ||
XM_005172286.4 | XP_005172343.1 | 4 - 5 | 561 | 67 | ||
XM_009303059.3 | XP_009301334.1 | 6 - 7 | 651 | 67 | ||
XM_009303060.3 | XP_009301335.1 | 5 - 6 | 585 | 67 | ||
GenBank mRNA | BC095884.1 | - | 4 - 5 | 488 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
