Assay Details
Gene Symbol: fbxl18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030245.2 | NP_001025416.2 | 3 - 4 | 1962 | 92 | |
GenBank mRNA | BC097162.1 | - | 3 - 4 | 92 | ||
BC139647.1 | - | 3 - 4 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
