Assay Details
Gene Symbol: mep1a.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030281.2 | NP_001025452.1 | 2 - 3 | 98 | 107 | |
GenBank mRNA | BC085666.1 | - | 2 - 3 | 61 | 107 | |
BC115219.1 | - | 2 - 3 | 98 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
