Assay Details
Gene Symbol: mep1a.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030281.2 | NP_001025452.1 | 6 - 7 | 379 | 129 | |
GenBank mRNA | BC085666.1 | - | 6 - 7 | 342 | 129 | |
BC115219.1 | - | 6 - 7 | 379 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
