Assay Details
Gene Symbol: gpr157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034174.1 | NP_001029346.1 | 2 - 3 | 743 | 78 | |
GenBank mRNA | BC100046.1 | - | 2 - 3 | 743 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
