Assay Details
Gene Symbol: loxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043325.1 | NP_001036790.1 | 5 - 6 | 1372 | 84 | |
GenBank mRNA | DQ304647.1 | - | 5 - 6 | 1372 | 84 | |
EF030480.1 | - | 5 - 6 | 1360 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
