Assay Details
Gene Symbol: tnfaip8l2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045301.1 | NP_001038766.1 | 1 - 2 | 417 | 63 | |
GenBank mRNA | BC117652.1 | - | 1 - 2 | 417 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
