Assay Details
Gene Symbol: ache | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131846.2 | NP_571921.1 | 3 - 4 | 1875 | 92 | |
GenBank mRNA | BC163891.1 | - | 3 - 4 | 1875 | 92 | |
BC163898.1 | - | 3 - 4 | 1875 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
