Assay Details
Gene Symbol: rpl7l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199590.1 | NP_955884.1 | 2 - 3 | 156 | 102 | |
GenBank mRNA | BC057532.1 | - | 2 - 3 | 156 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
