Assay Details
Gene Symbol: ssr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200053.1 | NP_956347.1 | 3 - 4 | 361 | 129 | |
GenBank mRNA | BC047859.1 | - | 3 - 4 | 361 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
