Assay Details
Gene Symbol: ythdf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200250.1 | NP_956544.1 | 2 - 3 | 195 | 84 | |
XM_005158763.4 | XP_005158820.1 | 2 - 3 | 224 | 84 | ||
GenBank mRNA | BC047846.1 | - | 2 - 3 | 195 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
