Assay Details
Gene Symbol: taf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200286.1 | NP_956580.1 | 4 - 5 | 444 | 81 | |
XM_005162303.4 | XP_005162360.1 | 4 - 5 | 534 | 81 | ||
GenBank mRNA | BC049422.1 | - | 4 - 5 | 444 | 81 | |
BC065457.1 | - | 4 - 5 | 481 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
