Assay Details
Gene Symbol: taf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200286.1 | NP_956580.1 | 7 - 8 | 739 | 101 | |
XM_005162303.4 | XP_005162360.1 | 7 - 8 | 949 | 221 | ||
GenBank mRNA | BC049422.1 | - | 7 - 8 | 739 | 101 | |
BC065457.1 | - | 7 - 8 | 896 | 221 |
Gene Symbol: LOC101885875 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009297018.3 | XP_009295293.1 | 2 - 3 | 302 | 221 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
