Assay Details
Gene Symbol: rpl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200713.1 | NP_957007.1 | 2 - 3 | 297 | 104 | |
XM_005167684.1 | XP_005167741.1 | 3 - 4 | 868 | 104 | ||
GenBank mRNA | BC059473.1 | - | 2 - 3 | 297 | 104 | |
BC065432.1 | - | 2 - 3 | 294 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
