Assay Details
Gene Symbol: gabra6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200731.1 | NP_957025.1 | 8 - 9 | 1020 | 139 | |
GenBank mRNA | BC059508.1 | - | 8 - 9 | 1020 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
