Assay Details
Gene Symbol: paip2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200766.1 | NP_957060.1 | 2 - 3 | 254 | 67 | |
GenBank mRNA | BC059597.1 | - | 2 - 3 | 254 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
