Assay Details
Gene Symbol: unc119b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040394.2 | NP_001035484.1 | 3 - 4 | 516 | 91 | |
GenBank mRNA | AF387341.1 | - | 3 - 4 | 355 | 91 | |
BC074025.1 | - | 3 - 4 | 441 | 91 | ||
BC094960.1 | - | 3 - 4 | 430 | 91 | ||
BC115147.1 | - | 3 - 4 | 650 | 91 | ||
BC124479.1 | - | 3 - 4 | 516 | 91 | ||
BC164428.1 | - | 3 - 4 | 540 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
