Assay Details
Gene Symbol: unc119b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040394.2 | NP_001035484.1 | 4 - 5 | 690 | 80 | |
GenBank mRNA | AF387341.1 | - | 4 - 5 | 529 | 80 | |
BC074025.1 | - | 4 - 5 | 615 | 80 | ||
BC094960.1 | - | 4 - 5 | 604 | 80 | ||
BC115147.1 | - | 4 - 5 | 824 | 80 | ||
BC124479.1 | - | 4 - 5 | 690 | 80 | ||
BC129251.1 | - | 4 - 5 | 803 | 80 | ||
BC164428.1 | - | 4 - 5 | 714 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
