Assay Details
Gene Symbol: dnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212795.1 | NP_997960.1 | 2 - 3 | 209 | 76 | |
GenBank mRNA | AY225448.1 | - | 2 - 3 | 209 | 76 | |
BC095578.1 | - | 2 - 3 | 171 | 76 | ||
BC164513.1 | - | 2 - 3 | 180 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
