Assay Details
Gene Symbol: egln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213310.1 | NP_998475.1 | 1 - 2 | 442 | 76 | |
GenBank mRNA | BC066699.1 | - | 1 - 2 | 442 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
