Assay Details
Gene Symbol: egln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213310.1 | NP_998475.1 | 3 - 4 | 700 | 110 | |
XM_005169814.4 | XP_005169871.1 | 3 - 4 | 788 | 110 | ||
GenBank mRNA | BC066699.1 | - | 3 - 4 | 700 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
