Assay Details
Gene Symbol: polr2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002317.2 | NP_001002317.2 | 3 - 4 | 453 | 74 | |
GenBank mRNA | AY648795.1 | - | 3 - 4 | 433 | 74 | |
BC095261.1 | - | 3 - 4 | 347 | 74 | ||
BC164278.1 | - | 3 - 4 | 378 | 74 | ||
EH541343.1 | - | 3 - 4 | 493 | 74 | ||
EH555518.1 | - | 3 - 4 | 490 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
