Assay Details
Gene Symbol: sec63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002588.1 | NP_001002588.1 | 1 - 2 | 206 | 91 | |
GenBank mRNA | BC076198.1 | - | 1 - 2 | 206 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
