Assay Details
Gene Symbol: src | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003837.2 | NP_001003837.2 | 8 - 9 | 1431 | 81 | |
XM_005174130.4 | XP_005174187.1 | 9 - 10 | 1489 | 81 | ||
XM_017353519.2 | XP_017209008.1 | 10 - 11 | 1517 | 81 | ||
XM_017353520.2 | XP_017209009.1 | 9 - 10 | 1499 | 81 | ||
GenBank mRNA | BC116544.1 | - | 8 - 9 | 1431 | 81 | |
BC165380.1 | - | 7 - 8 | 1070 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
