Assay Details
Gene Symbol: pgls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004117.3 | NP_001004117.2 | 4 - 5 | 552 | 130 | |
GenBank mRNA | BC080227.1 | - | 4 - 5 | 540 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
