Assay Details
Gene Symbol: clvs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025545.2 | NP_001020716.1 | 4 - 5 | 1325 | 82 | |
GenBank mRNA | BC116475.1 | - | 4 - 5 | 1325 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
