Assay Details
Gene Symbol: baiap2l2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080804.1 | NP_001074273.1 | 12 - 13 | 1434 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
