Assay Details
Gene Symbol: baiap2l2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080804.1 | NP_001074273.1 | 1 - 2 | 55 | 139 | |
XM_005161808.4 | XP_005161865.1 | 2 - 3 | 359 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
