Assay Details
Gene Symbol: naxd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110120.1 | NP_001103590.1 | 3 - 4 | 653 | 98 | |
NM_001328150.1 | NP_001315079.1 | 3 - 4 | 1643 | 98 | ||
XM_005167456.4 | XP_005167513.1 | 3 - 4 | 1622 | 98 | ||
XM_005167457.4 | XP_005167514.1 | 3 - 4 | 1604 | 98 | ||
XM_005167459.2 | XP_005167516.1 | 3 - 4 | 711 | 98 | ||
XM_005167460.4 | XP_005167517.1 | 3 - 4 | 989 | 98 | ||
XM_005167462.4 | XP_005167519.1 | 3 - 4 | 956 | 98 | ||
XM_005167463.4 | XP_005167520.1 | 2 - 3 | 672 | 98 | ||
XM_005167464.4 | XP_005167521.1 | 2 - 3 | 527 | 98 | ||
GenBank mRNA | BC153915.1 | - | 3 - 4 | 653 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
