Assay Details
Gene Symbol: col2a1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131292.1 | NP_571367.1 | 22 - 23 | 1472 | 108 | |
XM_005166863.4 | XP_005166920.1 | 21 - 22 | 1404 | 108 | ||
GenBank mRNA | BC163934.1 | - | 22 - 23 | 1464 | 108 | |
BC167477.1 | - | 21 - 22 | 1368 | 108 | ||
BC168504.1 | - | 22 - 23 | 1553 | 108 | ||
DQ335127.1 | - | 22 - 23 | 1472 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
