Assay Details
Gene Symbol: col2a1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131292.1 | NP_571367.1 | 43 - 44 | 3062 | 59 | |
XM_005166863.4 | XP_005166920.1 | 42 - 43 | 2994 | 59 | ||
GenBank mRNA | BC163934.1 | - | 43 - 44 | 3054 | 59 | |
BC167477.1 | - | 42 - 43 | 2958 | 59 | ||
BC168504.1 | - | 43 - 44 | 3143 | 59 | ||
DQ335127.1 | - | 43 - 44 | 3062 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
