Assay Details
Gene Symbol: synj2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199917.1 | NP_956211.1 | 3 - 4 | 404 | 98 | |
GenBank mRNA | BC045991.1 | - | 3 - 4 | 404 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
