Assay Details
Gene Symbol: rpa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200227.1 | NP_956521.1 | 3 - 4 | 348 | 122 | |
GenBank mRNA | BC046064.1 | - | 3 - 4 | 348 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
