Assay Details
Gene Symbol: tmem208 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207100.2 | NP_996983.1 | 878 | 153 | ||
GenBank mRNA | BC066516.1 | - | 4 - 5 | 446 | 153 | |
BC124418.1 | - | 878 | 153 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
