Assay Details
Gene Symbol: amy2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213011.2 | NP_998176.2 | 1 - 2 | 190 | 70 | |
GenBank mRNA | BC054669.1 | - | 1 - 2 | 165 | 70 | |
BC062867.1 | - | 1 - 2 | 167 | 70 | ||
CF103228.1 | - | 1 - 2 | 217 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
