Assay Details
Gene Symbol: grtp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213402.1 | NP_998567.1 | 1 - 2 | 259 | 84 | |
GenBank mRNA | BC059581.1 | - | 1 - 2 | 259 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
