Assay Details
Gene Symbol: grtp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213402.1 | NP_998567.1 | 4 - 5 | 696 | 115 | |
XM_021470179.1 | XP_021325854.1 | 4 - 5 | 455 | 115 | ||
GenBank mRNA | BC059581.1 | - | 4 - 5 | 696 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
