Assay Details
Gene Symbol: fgf17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214808.1 | NP_999973.1 | 3 - 4 | 439 | 92 | |
XM_005172491.4 | XP_005172548.1 | 3 - 4 | 869 | 92 | ||
XM_005172492.4 | XP_005172549.1 | 3 - 4 | 866 | 92 | ||
GenBank mRNA | AY457142.1 | - | 3 - 4 | 439 | 92 | |
BC083269.1 | - | 3 - 4 | 314 | 92 | ||
BC165029.1 | - | 3 - 4 | 284 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
