Assay Details
Gene Symbol: ccm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002315.3 | NP_001002315.1 | 2 - 3 | 501 | 107 | |
GenBank mRNA | AY648715.1 | - | 2 - 3 | 203 | 107 | |
BC092812.1 | - | 2 - 3 | 308 | 107 | ||
BC164183.1 | - | 2 - 3 | 237 | 107 | ||
DQ677878.1 | - | 2 - 3 | 236 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
