Assay Details
Gene Symbol: rdh12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002325.1 | NP_001002325.1 | 1 - 2 | 90 | 73 | |
GenBank mRNA | BC076473.1 | - | 1 - 2 | 90 | 73 | |
BC163997.1 | - | 1 - 2 | 107 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
