Assay Details
Gene Symbol: nedd8l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002557.1 | NP_001002557.1 | 3 - 4 | 266 | 94 | |
GenBank mRNA | BC076245.1 | - | 3 - 4 | 266 | 94 | |
BC151956.1 | - | 3 - 4 | 287 | 94 | ||
EF178472.1 | - | 3 - 4 | 321 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
