Assay Details
Gene Symbol: prkab1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002632.1 | NP_001002632.1 | 7 - 8 | 837 | 118 | |
XM_005155628.4 | XP_005155685.1 | 7 - 8 | 904 | 118 | ||
GenBank mRNA | BC075947.1 | - | 7 - 8 | 837 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
