Assay Details
Gene Symbol: glmp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004580.1 | NP_001004580.1 | 1 - 2 | 201 | 59 | |
GenBank mRNA | BC081645.1 | - | 1 - 2 | 201 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
