Assay Details
Gene Symbol: rtkn2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328566.1 | NP_001315495.1 | 3 - 4 | 114 | ||
GenBank mRNA | BC081383.1 | - | 3 - 4 | 623 | 114 | |
EH585343.1 | - | 3 - 4 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
