Assay Details
Gene Symbol: rtkn2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328566.1 | NP_001315495.1 | 5 - 6 | 753 | 70 | |
GenBank mRNA | BC081383.1 | - | 5 - 6 | 799 | 70 | |
EH585343.1 | - | 5 - 6 | 749 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
