Assay Details
Gene Symbol: c1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007059.1 | NP_001007060.1 | 2 - 3 | 173 | 114 | |
XM_021480568.1 | XP_021336243.1 | 2 - 3 | 182 | 114 | ||
GenBank mRNA | BC083407.1 | - | 2 - 3 | 173 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
